Gene Transcripts

Gene Symbol
Prf1
Entrez Gene ID
50669
Gene Name
perforin 1 (pore forming protein)
Gene Aliases
Cyta, RATCYTA
Chromosome Location
Chr. 20: 30915294 - 30920804 on Build Rnor_6.0
UniGene ID
Rn.11206
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017330.2 NP_059026.2 2 478
GenBank mRNA BC089808.1 - 2 478
M33605.1 - 1 426

View Full Product Details