Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000388.3 | NP_000379.2 | 4 | 1528 | |
NM_001178065.1 | NP_001171536.1 | 4 | 1594 | ||
XM_005247837.2 | XP_005247894.1 | 2 | 758 | ||
XM_006713789.3 | XP_006713852.1 | 4 | 2109 | ||
XM_017007324.1 | XP_016862813.1 | 5 | 3474 | ||
XM_017007325.1 | XP_016862814.1 | 4 | 1500 | ||
GenBank mRNA | BC104999.1 | - | 3 | 1180 | |
BC112236.1 | - | 3 | 1180 | 8327704 | |
D50855.1 | - | 3 | 1184 | ||
S83176.1 | - | 3 | 1156 | ||
U20759.1 | - | 4 | 1528 | ||
U20760.1 | - | 4 | 1594 |