Gene Transcripts

Gene Symbol
OR5I1
Entrez Gene ID
10798
Gene Name
olfactory receptor family 5 subfamily I member 1
Gene Aliases
HSOlf1, OLF1
Chromosome Location
Chr. 11: 55935456 - 55936400 on Build GRCh38
UniGene ID
Hs.533706
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006637.1 NP_006628.1 1 163
GenBank mRNA BC069093.1 - 1 163

View Full Product Details