Gene Transcripts

Gene Symbol
OR10J1
Entrez Gene ID
26476
Gene Name
olfactory receptor family 10 subfamily J member 1
Gene Aliases
HGMP07J, HSHGMP07J
Chromosome Location
Chr. 1: 159406348 - 159440967 on Build GRCh38
UniGene ID
Hs.532661
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012351.2 NP_036483.2 1 124
XM_006711271.2 XP_006711334.1 2 136
XM_017001015.1 XP_016856504.1 2 429
GenBank mRNA BC069150.1 - 1 87
BC112290.1 - 1 136 8327758
BC112292.1 - 1 124

View Full Product Details