Gene Transcripts

Gene Symbol
PNMA3
Entrez Gene ID
29944
Gene Name
paraneoplastic Ma antigen 3
Gene Aliases
MA3, MA5
Chromosome Location
Chr. X: 153056343 - 153060467 on Build GRCh38
UniGene ID
Hs.449627
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282535.1 NP_001269464.1 3 1655
NM_013364.5 NP_037496.4 2 1776
GenBank mRNA AF083116.1 - 1 1467
AL136878.1 - 3 1547

View Full Product Details