Gene Transcripts

Gene Symbol
SERTAD3
Entrez Gene ID
29946
Gene Name
SERTA domain containing 3
Gene Aliases
RBT1
Chromosome Location
Chr. 19: 40440841 - 40444375 on Build GRCh38
UniGene ID
Hs.515412
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013368.3 NP_037500.2 2 1066
NM_203344.2 NP_976219.1 2 955
XM_005258832.3 XP_005258889.1 3 1163
XM_006723179.3 XP_006723242.1 2 1015
GenBank mRNA AF192529.1 - 2 1079
AK074604.1 - 2 880
BC014061.2 - 2 879 3926937
BC050643.2 - 2 955 6503515

View Full Product Details