Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001081003.2 | NP_001074472.1 | 2 | 899 | |
NM_001081004.2 | NP_001074473.1 | 2 | 767 | ||
NM_001287237.1 | NP_001274166.1 | 2 | 1034 | ||
NM_014066.3 | NP_054785.2 | 2 | 921 | ||
GenBank mRNA | AF113540.1 | - | 2 | 1049 | |
AF290195.1 | - | 2 | 929 | ||
AK001477.1 | - | 2 | 747 | ||
AK023070.1 | - | 2 | 868 | ||
BC002672.2 | - | 2 | 751 | 3606279 | |
BC003055.1 | - | 2 | 839 | 3544106 | |
BC065729.1 | - | 2 | 889 | 6452461 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | XM_017008884.1 | XP_016864373.1 | 2 | 782 |