Gene Transcripts

Gene Symbol
HSPA4L
Entrez Gene ID
22824
Gene Name
heat shock protein family A (Hsp70) member 4 like
Gene Aliases
APG-1, HSPH3, Osp94
Chromosome Location
Chr. 4: 127781821 - 127834073 on Build GRCh38
UniGene ID
Hs.135554
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001317381.1 NP_001304310.1 4 597
NM_001317382.1 NP_001304311.1 3 316
NM_001317383.1 NP_001304312.1 3 607
NM_014278.3 NP_055093.2 3 607
XM_011531745.2 XP_011530047.1 3 676
GenBank mRNA AB023421.1 - 3 319
AK027822.1 - 3 494
AK302115.1 - 3 290
AK303053.1 - 4 597
AK307464.1 - 3 494
AY290862.1 - 3 473
BC040560.1 - 3 518 5270824
DB086945.1 - 3 316
EF197155.1 - 3 265

View Full Product Details