Gene Transcripts

Gene Symbol
GPR84
Entrez Gene ID
53831
Gene Name
G protein-coupled receptor 84
Gene Aliases
EX33, GPCR4
Chromosome Location
Chr. 12: 54362445 - 54365224 on Build GRCh38
UniGene ID
Hs.306199
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020370.2 NP_065103.1 2 685
XM_011538495.2 XP_011536797.1 2 1372
GenBank mRNA AF237762.1 - 2 651
AF282693.1 - 2 673
AK290394.1 - 1 1219
BC020614.1 - 2 683 4279185
FJ348261.1 - 1 583

View Full Product Details