Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004044.6 | NP_004035.2 | 3 | 359 | |
XM_017004187.1 | XP_016859676.1 | 3 | 282 | ||
GenBank mRNA | AB062403.1 | - | 3 | 207 | |
AK290067.1 | - | 2 | 341 | ||
AK298142.1 | - | 3 | 472 | ||
AK313066.1 | - | 3 | 253 | ||
BC008879.2 | - | 3 | 202 | 4300570 | |
D82348.1 | - | 3 | 247 | ||
D89976.1 | - | 3 | 229 | ||
DA043224.1 | - | 3 | 360 | ||
DA074758.1 | - | 3 | 346 | ||
EU794654.1 | - | 3 | 346 | ||
U37436.1 | - | 2 | 169 |