Gene Transcripts

Gene Symbol
MS4A6A
Entrez Gene ID
64231
Gene Name
membrane spanning 4-domains A6A
Gene Aliases
4SPAN3, 4SPAN3.2, CD20L3, CDA01, MS4A6, MST090, MSTP090
Chromosome Location
Chr. 11: 60171607 - 60184666 on Build GRCh38
UniGene ID
Hs.523702
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001247999.1 NP_001234928.1 3 695
NM_022349.3 NP_071744.2 2 506
NM_152851.2 NP_690590.1 2 506
NM_152852.2 NP_690591.1 3 631
XM_005274177.2 XP_005274234.1 3 690
XM_006718658.2 XP_006718721.1 3 565
XM_011545209.1 XP_011543511.1 3 565
XM_017018125.1 XP_016873614.1 3 565
GenBank mRNA AB013104.1 - 2 214
AF142409.1 - 2 234
AF172819.1 - 3 652
AF212240.1 - 2 375
AF253977.1 - 2 376
AF261136.1 - 2 373
AF350502.1 - 2 223
AF350503.1 - 2 223
AK291870.1 - 3 631
AK300725.1 - 2 240
BC022854.1 - 2 245 3934920
BC108728.1 - 2 279 6196213
DA917243.1 - 2 330
DA979138.1 - 2 277

View Full Product Details