Gene Transcripts

Gene Symbol
OR7A17
Entrez Gene ID
26333
Gene Name
olfactory receptor family 7 subfamily A member 17
Gene Aliases
BC85395_4, HTPCRX19
Chromosome Location
Chr. 19: 14880426 - 14881355 on Build GRCh38
UniGene ID
Hs.247717
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030901.1 NP_112163.1 1 487
GenBank mRNA BC101587.1 - 1 584
BC117357.1 - 1 584 40125908
X64993.1 - 1 113

View Full Product Details