Gene Transcripts

Gene Symbol
MTNR1B
Entrez Gene ID
4544
Gene Name
melatonin receptor 1B
Gene Aliases
FGQTL2, MEL-1B-R, MT2
Chromosome Location
Chr. 11: 92969623 - 92986241 on Build GRCh38
UniGene ID
Hs.569039
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005959.3 NP_005950.1 2 1218
XM_011542839.2 XP_011541141.1 2 1115
XM_017017777.1 XP_016873266.1 2 1521
GenBank mRNA BC069163.1 - 2 1218

View Full Product Details