Gene Transcripts
- Gene Symbol
- POLD2
- Entrez Gene ID
- 5425
- Gene Name
- DAN polymerase delta 2, accessory subunit
- Gene Aliases
- Chromosome Location
- Chr. 7: 44114680 - 44123668 on Build GRCh38
- UniGene ID
- Hs.306791
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001127218.2 |
NP_001120690.1
|
8, 9 |
1301 |
|
NM_001256879.1 |
NP_001243808.1
|
9, 10 |
1662 |
|
NM_006230.3 |
NP_006221.2
|
8, 9 |
1128 |
|
XM_011515431.1 |
XP_011513733.1
|
8, 9 |
1124 |
|
GenBank mRNA |
AK292347.1 |
- |
9, 10 |
1662 |
|
AK298168.1 |
- |
8, 9 |
992 |
|
AK312294.1 |
- |
8, 9 |
1261 |
|
BC000459.2 |
- |
8, 9 |
1091 |
2822169 |
BC030001.2 |
- |
7, 8 |
1068 |
|
U21090.1 |
- |
8, 9 |
1090 |
|
View Full Product Details