Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001304949.1 | NP_001291878.1 | 5 | 1054 | |
NM_001304951.1 | NP_001291880.1 | 2 | 743 | ||
NM_001304952.1 | NP_001291881.1 | 5 | 975 | ||
NM_001304953.1 | NP_001291882.1 | 5 | 1273 | ||
NM_001304954.1 | NP_001291883.1 | 4 | 1072 | ||
NM_152465.2 | NP_689678.1 | 4 | 1008 | ||
XM_005257927.4 | XP_005257984.1 | 5 | 1244 | ||
XM_006721723.3 | XP_006721786.1 | 5 | 1241 | ||
XM_011524398.2 | XP_011522700.1 | 5 | 974 | ||
XM_011524399.2 | XP_011522701.1 | 4 | 889 | ||
XM_011524400.2 | XP_011522702.1 | 4 | 798 | ||
XM_011524401.1 | XP_011522703.1 | 3 | 799 | ||
XM_017024237.1 | XP_016879726.1 | 4 | 892 | ||
GenBank mRNA | AK301868.1 | - | 5 | 972 | |
BC029574.1 | - | 4 | 955 | 5267334 | |
BQ215318.1 | - | 2 | 583 | ||
CR984201.1 | - | 5 | 735 |