Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_007679.4 | NP_031705.3 | 1 | 883 | |
GenBank mRNA | AK053190.1 | - | 1 | 883 | |
AK149723.1 | - | 348 | |||
AK151958.1 | - | 1 | 884 | ||
AK156762.1 | - | 1 | 883 | ||
BC139293.1 | - | 1 | 883 | 8862315 | |
X61800.1 | - | 1 | 1159 |