Gene Transcripts

Gene Symbol
TSR3
Entrez Gene ID
115939
Gene Name
TSR3, acp transferase ribosome maturation factor
Gene Aliases
C16orf42
Chromosome Location
Chr. 16: 1349240 - 1351872 on Build GRCh38
UniGene ID
Hs.134846
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001410.2 NP_001001410.1 4 613
GenBank mRNA BC013943.1 - 4 617
BC016699.1 - 4 617 4097703
BC017325.2 - 4 597 4549940

View Full Product Details