Gene Transcripts

Gene Symbol
OR52M1
Entrez Gene ID
119772
Gene Name
olfactory receptor family 52 subfamily M member 1
Gene Aliases
OR11-11, OR52M1P, OR52M3P
Chromosome Location
Chr. 11: 4545191 - 4546144 on Build GRCh38
UniGene ID
Hs.553567
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004137.1 NP_001004137.1 1 224

View Full Product Details