Gene Transcripts

Gene Symbol
KRTAP21-2
Entrez Gene ID
337978
Gene Name
keratin associated protein 21-2
Gene Aliases
KAP21.2
Chromosome Location
Chr. 21: 30746951 - 30747202 on Build GRCh38
UniGene ID
Hs.553699
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181617.1 NP_853648.1 1 107
GenBank mRNA AB096959.1 - 1 139

View Full Product Details