Gene Transcripts
- Gene Symbol
- SNX9
- Entrez Gene ID
- 51429
- Gene Name
- sorting nexin 9
- Gene Aliases
- SDP1, SH3PX1, SH3PXD3A, WISP
- Chromosome Location
- Chr. 6: 157716046 - 157945077 on Build GRCh38
- UniGene ID
- Hs.191213
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_016224.4 |
NP_057308.1
|
8, 9 |
1098 |
|
XM_005267015.2 |
XP_005267072.1
|
10, 11 |
1176 |
|
XM_011535886.2 |
XP_011534188.1
|
5, 6 |
637 |
|
GenBank mRNA |
AB209736.1 |
- |
4, 5 |
429 |
|
AF001629.1 |
- |
8, 9 |
903 |
|
AF076957.1 |
- |
3, 4 |
318 |
|
AF121859.1 |
- |
8, 9 |
858 |
|
AF131214.1 |
- |
8, 9 |
877 |
|
AF172847.1 |
- |
8, 9 |
978 |
|
AK026637.1 |
- |
5, 6 |
643 |
|
AK127345.1 |
- |
5, 6 |
953 |
|
AK314360.1 |
- |
8, 9 |
932 |
|
BC005022.1 |
- |
8, 9 |
1005 |
3832234 |
View Full Product Details