Gene Transcripts

Gene Symbol
OSTC
Entrez Gene ID
58505
Gene Name
oligosaccharyltransferase complex non-catalytic subunit
Gene Aliases
DC2
Chromosome Location
Chr. 4: 108650585 - 108667822 on Build GRCh38
UniGene ID
Hs.445803
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001267817.1 NP_001254746.1 2 245
NM_001267818.1 NP_001254747.1 2 245
NM_021227.3 NP_067050.1 2 245
GenBank mRNA AF068297.1 - 2 208
AF161425.1 - 2 229
AF201937.1 - 2 236
AF343342.1 - 2 174
AK312185.1 - 2 189
AW081061.1 - 2 246
BC016321.1 - 2 233 4075646
BC054857.1 - 2 235 4093030
BM471860.1 - 173
BM541430.1 - 2 196
BM546028.1 - 2 235
BQ435510.1 - 245
CB110420.1 - 2 190
CN430159.1 - 2 183

View Full Product Details