Gene Transcripts

Gene Symbol
CAMSAP2
Entrez Gene ID
23271
Gene Name
calmodulin regulated spectrin associated protein family member 2
Gene Aliases
CAMSAP1L1
Chromosome Location
Chr. 1: 200738905 - 200860707 on Build GRCh38
UniGene ID
Hs.23585
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001297707.1 NP_001284636.1 14 3957
NM_001297708.1 NP_001284637.1 13 3876
NM_203459.2 NP_982284.1 13 3924
XM_005245041.3 XP_005245098.1 14 3766
XM_017000799.1 XP_016856288.1 13 4350
GenBank mRNA AB029001.2 - 13 3315
BC011385.1 - 3 389
BC056910.1 - 3 2468
BC065508.1 - 3 2275
BC125229.1 - 14 3736 40124603
BC125230.1 - 13 3655 40124604

View Full Product Details