Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_138477.2 | NP_612486.2 | 23 | 3203 | |
XM_005254176.4 | XP_005254233.1 | 23 | 3118 | ||
XM_011521270.2 | XP_011519572.1 | 23 | 3143 | ||
XM_011521271.2 | XP_011519573.1 | 23 | 3140 | ||
XM_011521274.2 | XP_011519576.1 | 20 | 2189 | ||
GenBank mRNA | AF525398.1 | - | 23 | 3200 | |
BC001092.2 | - | 15 | 1658 | ||
BC008333.1 | - | 15 | 1658 | ||
BC008334.1 | - | 15 | 1658 | ||
BC052568.1 | - | 23 | 3091 | ||
BC066640.1 | - | 23 | 3091 | 6577335 |