Gene Transcripts

Gene Symbol
GAS7
Entrez Gene ID
8522
Gene Name
growth arrest specific 7
Gene Aliases
MLL/GAS7
Chromosome Location
Chr. 17: 9910609 - 10198551 on Build GRCh38
UniGene ID
Hs.462214
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130831.1 NP_001124303.1 14 3238
NM_003644.2 NP_003635.2 10 2987
NM_201432.1 NP_958836.1 14 3395
NM_201433.1 NP_958839.1 14 3425
XM_005256831.3 XP_005256888.1 15 3336
XM_005256832.3 XP_005256889.1 14 3206
XM_005256833.3 XP_005256890.1 13 3313
XM_005256834.3 XP_005256891.1 13 3139
XM_005256835.3 XP_005256892.1 13 3083
XM_011524044.2 XP_011522346.1 14 3230
XM_017025239.1 XP_016880728.1 14 3259
XM_017025240.1 XP_016880729.1 13 3042
XM_017025241.1 XP_016880730.1 13 3018
XM_017025242.1 XP_016880731.1 13 3192
GenBank mRNA AB007854.1 - 14 3193
AY327406.1 - 14 3425
BC001152.1 - 14 3218 3353809
BC006454.1 - 14 3395 3547000

View Full Product Details