Gene Transcripts

Gene Symbol
VMO1
Entrez Gene ID
284013
Gene Name
vitelline membrane outer layer 1 homolog
Gene Aliases
ERGA6350, PRO21055
Chromosome Location
Chr. 17: 4785285 - 4786434 on Build GRCh38
UniGene ID
Hs.122561
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001144939.1 NP_001138411.1 3 783
NM_001144940.1 NP_001138412.1 3 727
NM_001144941.1 NP_001138413.1 2 631
NM_182566.2 NP_872372.1 3 747
GenBank mRNA AF521892.1 - 3 752
AY359099.1 - 3 677
BC038467.1 - 2 630
BI836994.1 - 2 631

View Full Product Details