Gene Transcripts

Gene Symbol
OR4K14
Entrez Gene ID
122740
Gene Name
olfactory receptor family 4 subfamily K member 14
Gene Aliases
OR14-18, OR14-22
Chromosome Location
Chr. 14: 20014261 - 20015193 on Build GRCh38
UniGene ID
Hs.626814
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004712.1 NP_001004712.1 1 297

View Full Product Details