Gene Transcripts

Gene Symbol
ZDHHC13
Entrez Gene ID
54503
Gene Name
zinc finger DHHC-type containing 13
Gene Aliases
HIP14L, HIP3RP
Chromosome Location
Chr. 11: 19117099 - 19176420 on Build GRCh38
UniGene ID
Hs.188569
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001483.2 NP_001001483.1 10 1115
NM_019028.2 NP_061901.2 11 1261
XM_005252996.2 XP_005253053.1 11 1233
XM_011520193.1 XP_011518495.1 9 958
XM_011520194.2 XP_011518496.1 11 1236
XM_011520195.1 XP_011518497.1 11 1171
GenBank mRNA AB024495.1 - 11 1209
AB097014.1 - 11 1259
AB097027.1 - 11 1259
AK001714.1 - 11 1259
AK001831.1 - 6 633
AK291348.1 - 11 1236
AL833714.1 - 7 711
BC036020.1 - 11 1246 5272445
BC050690.1 - 10 1115 6171322
BC056152.1 - 11 1261 6170701
BX648156.1 - 10 1105

View Full Product Details