Gene Transcripts
- Gene Symbol
- Clec4a
- Entrez Gene ID
- 474143
- Gene Name
- C-type lectin domain family 4, member A
- Gene Aliases
- Clecsf6, Dcir1
- Chromosome Location
- Chr. 4: 156050040 - 156067518 on Build Rnor_6.0
- UniGene ID
- Rn.24032
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AY363176.1 |
- |
4 |
459 |
|
View Full Product Details