Gene Transcripts

Gene Symbol
Itm2b
Entrez Gene ID
290364
Gene Name
integral membrane protein 2B
Gene Aliases
Bri, imBRI2
Chromosome Location
Chr. 15: 55254703 - 55277713 on Build Rnor_6.0
UniGene ID
Rn.107335
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001006963.1 NP_001006964.1 4 707
GenBank mRNA BC083735.1 - 4 707
FQ210227.1 - 4 737
FQ212539.1 - 1 84
FQ212925.1 - 4 664
FQ218718.1 - 4 706
FQ219378.1 - 1 70
FQ229426.1 - 4 706
FQ230065.1 - 4 679

View Full Product Details