Gene Transcripts

Gene Symbol
RESP18
Entrez Gene ID
389075
Gene Name
regulated endocrine specific protein 18
Gene Aliases
Chromosome Location
Chr. 2: 219327409 - 219336656 on Build GRCh38
UniGene ID
Hs.290551
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007089.3 NP_001007090.3 5, 6 554
XM_011511165.2 XP_011509467.1 3, 4 417
XM_011511167.2 XP_011509469.1 4, 5 868
XM_017004079.1 XP_016859568.1 4, 5 711
GenBank mRNA AF437883.3 - 5, 6 554
DQ227570.1 - 3, 4 625

View Full Product Details