Gene Transcripts

Gene Symbol
Ermn
Entrez Gene ID
295619
Gene Name
ermin, ERM-like protein
Gene Aliases
RGD1308367, juxtanodin
Chromosome Location
Chr. 3: 44080350 - 44086006 on Build Rnor_6.0
UniGene ID
Rn.20243
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008311.3 NP_001008312.1 3 954
GenBank mRNA BC086596.1 - 3 954
DQ119821.1 - 882

View Full Product Details