Gene Transcripts

Gene Symbol
Vom1r44
Entrez Gene ID
494254
Gene Name
vomeronasal 1 receptor 44
Gene Aliases
V1rg7
Chromosome Location
Chr. 1: 67174653 - 67181472 on Build Rnor_6.0
UniGene ID
Rn.129423
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008902.1 NP_001008902.1 1 850
XM_008758769.1 XP_008756991.1 3 1872
XM_008758770.1 XP_008756992.1 4 1394
GenBank mRNA AY510307.1 - 1 850

View Full Product Details