Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_014740.3 | NP_055555.1 | 3 | 489 | |
XM_011525522.1 | XP_011523824.1 | 3 | 445 | ||
GenBank mRNA | AK290608.1 | - | 3 | 470 | |
BC003662.2 | - | 3 | 399 | 2988931 | |
BC004386.1 | - | 3 | 420 | 3619386 | |
BC011151.1 | - | 3 | 424 | 3452439 | |
CR456750.1 | - | 3 | 267 | ||
D21853.1 | - | 3 | 481 | ||
DA667983.1 | - | 3 | 489 | ||
X79538.1 | - | 3 | 340 |