Gene Transcripts

Gene Symbol
Trim10
Entrez Gene ID
294210
Gene Name
tripartite motif-containing 10
Gene Aliases
Rfb30, Rnf9
Chromosome Location
Chr. 20: 2182945 - 2191640 on Build Rnor_6.0
UniGene ID
Rn.140858
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009176.2 NP_001009176.1 3 911
GenBank mRNA BC129078.1 - 3 911

View Full Product Details