Gene Transcripts

Gene Symbol
SNX17
Entrez Gene ID
9784
Gene Name
sorting nexin 17
Gene Aliases
Chromosome Location
Chr. 2: 27370496 - 27377533 on Build GRCh38
UniGene ID
Hs.278569
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001267059.1 NP_001253988.1 3 444
NM_001267060.1 NP_001253989.1 2 405
NM_001267061.1 NP_001253990.1 3 315
NM_014748.3 NP_055563.1 3 480
NR_049782.1 3 605
NR_049783.1 3 504
NR_049784.1 3 480
GenBank mRNA AK129689.1 - 3 364
AK222543.1 - 3 363
AK293252.1 - 3 380
AK293464.1 - 3 296
AK297054.1 - 3 489
AK298620.1 - 3 315
AK298869.1 - 2 266
AK300144.1 - 3 327
AK316373.1 - 2 185
BC002524.1 - 3 341 3140610
BC002610.1 - 3 330 3143104
BC014620.2 - 3 341 3010648
BC050590.1 - 3 335 6044270
BT007167.1 - 3 232
CR457081.1 - 3 232
D31764.1 - 3 454
DB177194.1 - 3 470

View Full Product Details