Gene Transcripts

Gene Symbol
Oas1e
Entrez Gene ID
494201
Gene Name
2'-5' oligoadenylate synthetase 1E
Gene Aliases
Oas1c
Chromosome Location
Chr. 12: 41281558 - 41290500 on Build Rnor_6.0
UniGene ID
Rn.144034
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009492.1 NP_001009492.1 6 1598
GenBank mRNA AY196698.1 - 6 1598

View Full Product Details