Gene Transcripts

Gene Symbol
Fam115e
Entrez Gene ID
494321
Gene Name
family with sequence similarity 115, member E
Gene Aliases
Eapa2
Chromosome Location
Chr. 4: 72010583 - 72023952 on Build Rnor_6.0
UniGene ID
Rn.108070
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009534.1 NP_001009534.1 2 551
GenBank mRNA BC061995.1 - 2 551

View Full Product Details