Gene Transcripts

Gene Symbol
Chic1
Entrez Gene ID
12212
Gene Name
cysteine-rich hydrophobic domain 1
Gene Aliases
Brx
Chromosome Location
Chr. X: 103356328 - 103409096 on Build GRCm38
UniGene ID
Mm.42223
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009767.2 NP_033897.1 2 434
GenBank mRNA AK034427.1 - 2 407
AK135974.1 - 2 386
BC132322.1 - 2 353 40130599
BC138730.1 - 2 353 8861752

View Full Product Details