Gene Transcripts

Gene Symbol
Clint1
Entrez Gene ID
360515
Gene Name
clathrin interactor 1
Gene Aliases
Clint, Enth, Epn4
Chromosome Location
Chr. 10: 30833914 - 31007813 on Build Rnor_6.0
UniGene ID
Rn.229329
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001002022.1 NP_001002022.1 10 1672
GenBank mRNA BC076397.1 - 10 1672

View Full Product Details