Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001003919.1 | NP_001003919.1 | 27 | 3369 | |
GenBank mRNA | AK076357.1 | - | 26 | 4169 | |
AK152868.1 | - | 27 | 3358 | ||
BC049929.1 | - | 15 | 1883 | ||
BC079656.1 | - | 27 | 3369 | 6854039 |