Gene Transcripts

Gene Symbol
TUSC1
Entrez Gene ID
286319
Gene Name
tumor suppressor candidate 1
Gene Aliases
TSG-9, TSG9
Chromosome Location
Chr. 9: 25676389 - 25678858 on Build GRCh38
UniGene ID
Hs.26268
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004125.2 NP_001004125.1 1 1839
GenBank mRNA AY168647.2 - 1 1421
BC017510.1 - 1 1214
BC028316.1 - 1 938
BC036931.1 - 1 917
BC046919.1 - 1 1280
BC110824.1 - 1 1421 5169320

View Full Product Details