Gene Transcripts
- Gene Symbol
- Nlrp1a
- Entrez Gene ID
- 195046
- Gene Name
- NLR family, pyrin domain containing 1A
- Gene Aliases
- CARD7, DEFCAP, Gm14, Gm15, NAC, Nalp1, Nalp1a, Nlrp1, PP1044
- Chromosome Location
- Chr. 11: 71091197 - 71144704 on Build GRCm38
- UniGene ID
- Mm.240227
- Species
- Mus musculus
- Gene Symbol
- Nlrp1c-ps
- Entrez Gene ID
- 627984
- Gene Name
- NLR family, pyrin domain containing 1C, pseudogene
- Gene Aliases
- Nalp1c, Nlrp1c, OTTMUSG00000006090
- Chromosome Location
- Chr. 11: 71242430 - 71285232 on Build GRCm38
- UniGene ID
- Mm.390378
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC141384.1 |
- |
2 |
1273 |
|
DQ117602.1 |
- |
2 |
1277 |
|
View Full Product Details