Gene Transcripts

Gene Symbol
Fscn3
Entrez Gene ID
296947
Gene Name
fascin actin-bundling protein 3, testicular
Gene Aliases
Chromosome Location
Chr. 4: 55720020 - 55729442 on Build Rnor_6.0
UniGene ID
Rn.3253
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004232.1 NP_001004232.1 2 871
GenBank mRNA BC079410.1 - 2 871

View Full Product Details