Gene Transcripts

Gene Symbol
MUC17
Entrez Gene ID
140453
Gene Name
mucin 17, cell surface associated
Gene Aliases
MUC-17, MUC-3, MUC3
Chromosome Location
Chr. 7: 101020083 - 101058859 on Build GRCh38
UniGene ID
Hs.271819
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001040105.1 NP_001035194.1 3 1029
NR_133665.1 3 1029
GenBank mRNA AJ606307.1 - 3 1029
AJ606308.1 - 3 1029

View Full Product Details