Gene Transcripts

Gene Symbol
NCAPD2
Entrez Gene ID
9918
Gene Name
non-SMC condensin I complex subunit D2
Gene Aliases
CAP-D2, CNAP1, hCAP-D2
Chromosome Location
Chr. 12: 6494132 - 6531966 on Build GRCh38
UniGene ID
Hs.5719
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014865.3 NP_055680.3 2 144
GenBank mRNA AK022511.1 - 2 144
AK125155.1 - 2 183
AK128354.1 - 2 173
AK303310.1 - 2 178
BC016913.1 - 2 876
BC028182.1 - 2 156 5395775
CN307514.1 - 1 122
D63880.1 - 2 897

View Full Product Details