Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001305792.1 | NP_001292721.1 | 1 | 295 | |
NM_014875.2 | NP_055690.1 | 2 | 619 | ||
XM_011510231.2 | XP_011508533.1 | 3 | 713 | ||
XM_011510232.2 | XP_011508534.1 | 2 | 375 | ||
XM_011510236.1 | XP_011508538.1 | 2 | 590 | ||
XM_017003005.1 | XP_016858494.1 | 1 | 796 | ||
XM_017003006.1 | XP_016858495.1 | 2 | 404 | ||
GenBank mRNA | BC113742.1 | - | 1 | 258 | |
D26361.1 | - | 2 | 619 |