Gene Transcripts

Gene Symbol
PUM3
Entrez Gene ID
9933
Gene Name
pumilio RNA binding family member 3
Gene Aliases
HA-8, HLA-HA8, KIAA0020, PEN, PUF-A, PUF6, XTP5
Chromosome Location
Chr. 9: 2804152 - 2844130 on Build GRCh38
UniGene ID
Hs.493309
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014878.4 NP_055693.4 8 907
GenBank mRNA AF490254.1 - 5 391
AK225953.1 - 8 872
AK292169.1 - 8 850
AK315531.1 - 8 886
AL832239.1 - 8 873
AL832245.1 - 8 866
AY047588.1 - 1 109
AY047589.1 - 1 109
AY047590.1 - 1 109
BC016137.2 - 7 811 3908516
CR456957.1 - 5 391
D13645.1 - 7 809

View Full Product Details