Gene Transcripts

Gene Symbol
Creb3l1
Entrez Gene ID
362165
Gene Name
cAMP responsive element binding protein 3-like 1
Gene Aliases
Oasis
Chromosome Location
Chr. 3: 80892427 - 80933339 on Build Rnor_6.0
UniGene ID
Rn.24541
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005562.1 NP_001005562.1 12 2453
XM_006234584.1 XP_006234646.1 12 2472
GenBank mRNA BC081951.1 - 12 2453

View Full Product Details