Gene Transcripts

Gene Symbol
Ghitm
Entrez Gene ID
290596
Gene Name
growth hormone inducible transmembrane protein
Gene Aliases
MICS1
Chromosome Location
Chr. 16: 14370269 - 14382647 on Build Rnor_6.0
UniGene ID
Rn.98375
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005908.1 NP_001005908.1 7 853
XM_006252786.2 XP_006252848.1 7 1107
GenBank mRNA BC083778.1 - 7 853
FQ212260.1 - 7 1053
FQ217853.1 - 7 871

View Full Product Details