Gene Transcripts

Gene Symbol
Leo1
Entrez Gene ID
235497
Gene Name
Leo1, Paf1/RNA polymerase II complex component, homolog (S. cerevisiae)
Gene Aliases
Gm185
Chromosome Location
Chr. 9: 75441524 - 75466432 on Build GRCm38
UniGene ID
Mm.41508
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BC082540.1 - 7 2624

View Full Product Details